missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals DL-Arabinose, 98+%

Product Code. 11362337
Change view
Click to view available options
Quantity:
10 g
Conditionnement:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantity unitSize
11362337 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 11362337 Fournisseur Thermo Scientific Chemicals Code fournisseur L09895.09
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

CAS: 147-81-9 | C5H10O5 | 150.13 g/mol

DL-Arabinose is used in physicochemical studies to measure properties such as dielectric relaxation. It may be used in chiral separation of sugars. DL-Arabinose, is a building block of polysaccharide (CCPa-1) from fungus Coprinus comatus Gray, which has potential application as natural antitumor agent with immunomodulatory activity. Arabinose is also used as a culture medium for certain bacteria.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-Arabinose is used in physicochemical studies to measure properties such as dielectric relaxation. It may be used in chiral separation of sugars. DL-Arabinose, is a building block of polysaccharide (CCPa-1) from fungus Coprinus comatus Gray, which has potential application as natural antitumor agent with immunomodulatory activity. Arabinose is also used as a culture medium for certain bacteria.

Solubility
Soluble in Water: 250 mg/5ml.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 147-81-9
Molecular Formula C5H10O5
Molecular Weight (g/mol) 150.13
MDL Number MFCD00135866,MFCD00135608,MFCD00135867,MFCD00135866
InChI Key SRBFZHDQGSBBOR-UHFFFAOYNA-N
Synonym l-+-arabinose, dl-arabinose, 2r,3s,4s-2,3,4,5-tetrahydroxypentanal, aldehydo-l-arabinose, arabinose, l, unii-b40roo395z, pectinose, pectin sugar, aldehydo-l-arabino-pentose, arabinose,l
PubChem CID 5460291
ChEBI CHEBI:6182
SMILES OC1COC(O)C(O)C1O

Spécification

Melting Point 158°C to 160°C
Quantity 10 g
Beilstein 1723086
Merck Index 14,761
Solubility Information Soluble in Water: 250mg/5ml.
IUPAC Name (2R,3S,4S)-2,3,4,5-tetrahydroxypentanal
Formula Weight 150.13
Percent Purity ≥98%
Chemical Name or Material DL-Arabinose

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.