Learn More
Thermo Scientific Chemicals DL-Homocysteinethiolactone hydrochloride, 99%
Product Code. 10152390 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10152390 1 g Glass bottle 1g
10633342 25 g Glass bottle 25g
10103111 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10152390 Supplier Thermo Scientific Chemicals Supplier No. 120830010
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
CAS: 6038-19-3 | C4H7NOS·HCl | 153.63 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 6038-19-3
MDL Number MFCD00012724
InChI Key ZSEGSUBKDDEALH-UHFFFAOYSA-N
Synonym dl-homocysteine thiolactone hydrochloride, 3-aminodihydrothiophen-2 3h-one hydrochloride, 3-aminothiolan-2-one hydrochloride, hctl hydrochloride, dl-homocysteinethiolactone hydrochloride, d,l-homocysteine thiolactone hydrochloride, d,l-homocysteinthiolakton chlorid german, +--dihydro-3-amino-2 3h-thiophenone hydrochloride, 2 3h-thiophenone, dihydro-3-amino-, hydrochloride, +-, l-2-amino-4-mercaptobutyric acid 1,4-thiolactone hydrochloride
PubChem CID 110753
IUPAC Name 3-aminothiolan-2-one;hydrochloride
SMILES C1CSC(=O)C1N.Cl
Specifications
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C4H8ClNOS
Quantity 1 g
Merck Index 15, 4772
Sublimation Point >140°C
Molecular Weight (g/mol) 153.624
Formula Weight 153.63
Percent Purity ≥98.5%
Physical Form Fine Crystalline Powder
Chemical Name or Material DL-Homocysteinethiolactone hydrochloride, 99%