Learn More
DL-Homophenylalanine, TRC
Product Code. 30135902
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135902 5 g 5g
17957154 250 mg 250mg
17988574 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30135902 Supplier Toronto Research Chemicals Supplier No. TRCH5913105G
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1012-05-1
Molecular Formula C10 H13 N O2
Molecular Weight (g/mol) 179.22
Synonym Benzenebutanoic acid, α-amino-, Benzenebutanoic acid, α-amino-, (±)-, Butyric acid, 2-amino-4-phenyl-, DL- (8CI), α-Aminobenzenebutanoic acid, 2-Amino-4-phenylbutanoic acid, 2-Amino-4-phenylbutyric acid, DL-2-Amino-4-phenylbutyric acid, DL-Homophenylalanine, (2RS)-2-Amino-4-phenylbutanoic acid
IUPAC Name 2-amino-4-phenylbutanoic acid
SMILES NC(CCc1ccccc1)C(=O)O
Specifications
InChI Formula InChI=1S/C10H13NO2/c11-9(10(12)13)7-6-8-4-2-1-3-5-8/h1-5,9H,6-7,11H2,(H,12,13)
Quantity 5 g
Formula Weight 179.0946
Shipping Condition Room Temperature
Chemical Name or Material (2RS)-2-Amino-4-phenylbutanoic Acid