missing translation for 'onlineSavingsMsg'
Learn More

DL-Laudanosine, TRC

Product Code. 18032900
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18032900 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18032900 Supplier Toronto Research Chemicals Supplier No. TRCL17854010MG
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1699-51-0
Molecular Formula C21 H27 N O4
Quantity 10 mg
Synonym 1-[(3,4-dimethoxyphenyl)methyl]-1,2,3,4-tetrahydro-6,7-dimethoxy-2-methyl-Isoquinoline, Isoquinoline, 1,2,3,4-tetrahydro-6,7-dimethoxy-2-methyl-1-veratryl-, (±)- (8CI), Isoquinoline, 1-[(3,4-dimethoxyphenyl)methyl]-1,2,3,4-tetrahydro-6,7-dimethoxy-2-methyl-, (±)-, (±)-Laudanosine, 1,2,3,4-Tetrahydro-6,7-dimethoxy-2-methyl-1-veratrylisoquinoline, DL-Laudanosine, N-Methyltetrahydropapaverine, NSC 331268, NSC 94267, Veraisoquin, Atracurium Besilate Imp. G (EP)
InChI Formula InChI=1S/C21H27NO4/c1-22-9-8-15-12-20(25-4)21(26-5)13-16(15)17(22)10-14-6-7-18(23-2)19(11-14)24-3/h6-7,11-13,17H,8-10H2,1-5H3
SMILES COc1ccc(CC2N(C)CCc3cc(OC)c(OC)cc23)cc1OC
IUPAC Name 1-[(3,4-dimethoxyphenyl)methyl]-6,7-dimethoxy-2-methyl-3,4-dihydro-1H-isoquinoline
Molecular Weight (g/mol) 357.44
Formula Weight 357.194
Percent Purity >95%
Chemical Name or Material Laudanosine
Grade HPLC
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.