missing translation for 'onlineSavingsMsg'
Learn More

DL-Norepinephrine Hydrochloride, TRC

Product Code. 30134517
Change view
Click to view available options
Quantity:
50 mg
100 mg
1 g
Unit Size:
100mg
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134517 50 mg 50mg
30040650 100 mg 100mg
18072740 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134517 Supplier Toronto Research Chemicals Supplier No. TRCN67450050MG
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55-27-6
Molecular Formula C8 H11 N O3 . Cl H
Molecular Weight (g/mol) 205.64
Synonym 1,2-Benzenediol, 4-(2-amino-1-hydroxyethyl)-, hydrochloride (1:1), 1,2-Benzenediol, 4-(2-amino-1-hydroxyethyl)-, hydrochloride (9CI), 1,2-Benzenediol, 4-(2-amino-1-hydroxyethyl)-, hydrochloride, (±)-, Benzyl alcohol, α-(aminomethyl)-3,4-dihydroxy-, hydrochloride, (±)- (8CI), (±)-Noradrenaline hydrochloride, (±)-Norepinephrine hydrochloride, DL-Arterenol hydrochloride, DL-Norepinephrine hydrochloride, NSC 7930, dl-Noradrenaline hydrochloride, dl-Norepinephrine hydrochloride
IUPAC Name 4-(2-amino-1-hydroxyethyl)benzene-1,2-diol;hydrochloride
SMILES Cl.NCC(O)c1ccc(O)c(O)c1

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C8H11NO3.ClH/c9-4-8(12)5-1-2-6(10)7(11)3-5;/h1-3,8,10-12H,4,9H2;1H
Formula Weight 205.0506
Percent Purity >95%
Chemical Name or Material (±)-Norepinephrine (Noradrenaline) Hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.