missing translation for 'onlineSavingsMsg'
Learn More

DL-p-Hydroxyphenyllactic Acid, TRC

Product Code. 30134384
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134384 500 mg 500mg
30133600 1 g 1g
17931505 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134384 Supplier Toronto Research Chemicals Supplier No. TRCH953665500MG
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 306-23-0
Molecular Formula C9 H10 O4
Molecular Weight (g/mol) 182.17
Synonym 3-(p-Hydroxyphenyl)lactic Acid, (RS)-3-(4-Hydroxyphenyl)lactic Acid, (±)-3-(4-Hydroxyphenyl)lactic Acid, 2-Hydroxy-3-(4-hydroxyphenyl)propanoic Acid, 2-Hydroxy-3-(p-hydroxyphenyl)propionic Acid, 3-(4-Hydroxyphenyl)-DL-lactic Acid, 3-(4-Hydroxyphenyl)lactic Acid, DL-3-(4-Hydroxyphenyl)lactic Acid, HPLA, NSC 111175, p-Hydroxy-dl-phenyllactic Acid, α,4-Dihydroxybenzenepropanoic Acid, β-(4-Hydroxyphenyl)lactic Acid, β-(p-Hydroxyphenyl)-DL-lactic Acid, β-(p-Hydroxyphenyl)lactic Acid, Lactic acid, 3-(p-hydroxyphenyl)- (6CI,8CI)
IUPAC Name 2-hydroxy-3-(4-hydroxyphenyl)propanoic acid
SMILES OC(Cc1ccc(O)cc1)C(=O)O

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C9H10O4/c10-7-3-1-6(2-4-7)5-8(11)9(12)13/h1-4,8,10-11H,5H2,(H,12,13)
Formula Weight 182.0579
Chemical Name or Material DL-p-Hydroxyphenyllactic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.