missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Frataxin Antibody (OTI1C12), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70766AF594
This item is not returnable.
View return policy
Description
Frataxin Monoclonal specifically detects Frataxin in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| Frataxin | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| FXN | |
| Full length recombinant protein of human Frataxin (NP_000135) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI1C12 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| CyaY, EC 1.16.3.1, FA, FARRmitochondrial, frataxin, Friedreich ataxia, Friedreich ataxia protein, Fxn, X25MGC57199 | |
| Mouse | |
| Affinity Purified | |
| Diabetes Research | |
| 2395.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction