missing translation for 'onlineSavingsMsg'
Learn More

DMDM Hydantoin, TRC

Product Code. 30134543
Change view
Click to view available options
Quantity:
1 g
2.5 g
5 g
Unit Size:
1g
2.5g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134543 5 g 5g
17974685 1 g 1g
17912205 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134543 Supplier Toronto Research Chemicals Supplier No. TRCP6881355G
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6440-58-0
Molecular Formula C7 H12 N2 O4
Molecular Weight (g/mol) 188.18
Synonym 1,3-bis(Hydroxymethyl)-5,5-dimethylhydantoin, DMDM Hydantoin, 1,3-Bis(hydroxymethyl)-5,5-dimethylhydantoin, 1,3-Bis(hydroxymethyl)-5,5-dimethylimidazolidine-2,4-dione, 1,3-Dihydroxymethyl-5,5-dimethylhydantoin, 1,3-Dimethylol-5,5-dimethylhydantoin, DMDMH
IUPAC Name 1,3-bis(hydroxymethyl)-5,5-dimethylimidazolidine-2,4-dione
SMILES CC1(C)N(CO)C(=O)N(CO)C1=O

Specifications

Quantity 5 g
InChI Formula InChI=1S/C7H12N2O4/c1-7(2)5(12)8(3-10)6(13)9(7)4-11/h10-11H,3-4H2,1-2H3
Formula Weight 188.0797
Percent Purity >95%
Chemical Name or Material DMDM Hydantoin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.