missing translation for 'onlineSavingsMsg'
Learn More

DNA-PKcs Ab-1 Monoclonal Antibody, Epredia™

Inhibit DNA-dependent phosphorylation and perform other studies with Epredia™ DNA-PKcs Ab-1, Mouse Monoclonal Antibody.

Brand:  Epredia MS-369-P

Additional Details : Weight : 0.00100kg

 View more versions of this product

Product Code. 12641197

  • 750.00€ / 1mL
Estimated Shipment: 18-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

DNA-dependent Protein Kinase (DNA-PKcs) phosphorylates transcription factors, including Sp1, Oct-1, p53, and SV40 large T antigen in vitro. In vivo, it is involved in repairing double strand DNA breaks.

Host Species: Mouse

Clone: 18-2

Isotype: IgG1

Species Reactivity: Human and Rat. Others not known.

Epitope: aa 1-2713

Immunogen: Human DNA-dependent Protein Kinase purified from HeLa cells

Molecular Weight: 460 kDa

Positive Control: LS174T cells, Tonsil

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native and denatured)
  • Inhibits (50%) DNA-dependent Phosphorylation

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

DNA-PKcs Ab-1
Monoclonal
200μg/mL
Mouse
1 mL
DNA Repair
Human, Rat
IgG1
Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
18-2
Unconjugated
Human DNA-dependent Protein Kinase purified from HeLa cells
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions