Learn More
DNA-PKcs Ab-1 Monoclonal Antibody, Epredia™
Inhibit DNA-dependent phosphorylation and perform other studies with Epredia™ DNA-PKcs Ab-1, Mouse Monoclonal Antibody.
Brand: Epredia MS-369-P1ABX
Additional Details : Weight : 1.00000kg
Description
DNA-dependent Protein Kinase (DNA-PKcs) phosphorylates transcription factors, including Sp1, Oct-1, p53, and SV40 large T antigen in vitro. In vivo, it is involved in repairing double strand DNA breaks.Host Species: Mouse
Clone: 18-2
Isotype: IgG1
Species Reactivity: Human and Rat. Others not known.
Epitope: aa 1-2713
Immunogen: Human DNA-dependent Protein Kinase purified from HeLa cells
Molecular Weight: 460 kDa
Positive Control: LS174T cells, Tonsil
Cellular Localization: Nuclear
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
- Immunoprecipitation (Native and denatured)
- Inhibits (50%) DNA-dependent Phosphorylation
Note:
USA: RUO; Int'l: RUO
Specifications
DNA-PKcs Ab-1 | |
Monoclonal | |
1mg/mL | |
Mouse | |
100 μL | |
DNA Repair | |
Human, Rat | |
IgG1 |
Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
18-2 | |
Unconjugated | |
Human DNA-dependent Protein Kinase purified from HeLa cells | |
RUO | |
Primary | |
Concentrated |