missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DNAH9 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA545744
This item is not returnable.
View return policy
Description
Peptide sequence: DSQARDGAGA TREEKVKALL EEILERVTDE FNIPELMAKV EERTPYIVVA Sequence homology: Cow: 83%; Dog: 100%; Guinea Pig: 83%; Horse: 92%; Human: 100%; Mouse: 92%; Rat: 92%.
This gene encodes the heavy chain subunit of axonemal dynein, a large multi-subunit molecular motor. Axonemal dynein attaches to microtubules and hydrolyzes ATP to mediate the movement of cilia and flagella. The gene expresses at least two transcript variants; additional variants have been described, but their full length nature has not been determined.
Specifications
| DNAH9 | |
| Polyclonal | |
| Unconjugated | |
| DNAH9 | |
| 9030022M04; A230091C01; axonemal beta dynein heavy chain 9; Bv2; C230051G16; ciliary dynein heavy chain 9; D11Ertd686e; DNAH17L; Dnah9; DNAH9 variant protein; Dnahc9; DNAL1; DNEL1; DYH9; dynein axonemal heavy chain 9; dynein heavy chain; dynein heavy chain 9, axonemal; dynein heavy chain Bv2; dynein, axonemal, heavy chain 9; dynein, axonemal, heavy polypeptide 9; HL20; HL-20; KIAA0357; mKIAA0357 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 1770, 237806 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9NYC9 | |
| DNAH9 | |
| Synthetic peptide directed towards the C-terminal of human DNAH9 (aa 4206-4255). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction