missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNAJB14 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16501-1-AP-20UL
This item is not returnable.
View return policy
Specifications
| DNAJB14 | |
| Polyclonal | |
| Unconjugated | |
| DNAJB14 | |
| DNAJB14, EGNR9427, PRO34683 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 499716, 70604, 79982 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot, Immunofluorescence, Immunocytochemistry | |
| 0.32 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q149L6, Q8TBM8 | |
| Dnajb14 | |
| DNAJB14 Fusion Protein Ag9668 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction