missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNAJC10 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13101-1-AP-20UL
This item is not returnable.
View return policy
Description
The ERdj5 antibody displays a higher affinity for endogenous ERdj5u.
This endoplasmic reticulum co-chaperone may play a role in protein folding and translocation across the endoplasmic reticulum membrane. DNAJC10 may act as a co-chaperone for HSPA5.Specifications
| DNAJC10 | |
| Polyclonal | |
| Unconjugated | |
| DNAJC10 | |
| DNAJC10, ER resident protein ERdj5, ERdj5, JPDI, Macrothioredoxin, MTHr | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 295690, 54431, 66861 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.28 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q498R3, Q8IXB1, Q9DC23 | |
| DNAJC10 | |
| DNAJC10 Fusion Protein Ag3737 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur