missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNAJC12 Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-33236-100ul
This item is not returnable.
View return policy
Description
DNAJC12 Monoclonal antibody specifically detects DNAJC12 in Mouse samples. It is validated for ELISA,Western Blot
Specifications
| DNAJC12 | |
| Monoclonal | |
| Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL | |
| DnaJ (Hsp40) homolog, subfamily C, member 12, dnaJ homolog subfamily C member 12, J domain containing protein 1 (JDP1), J domain protein 1, JDP1J domain-containing protein 1 | |
| A synthetic peptide corresponding to a sequence within amino acids 99-198 of human DNAJC12 (Q9UKB3).,, Sequence:, TSMHWVVRGKKDLMLEESDKTHTTKMENEECNEQRERKKEELASTAEKTEQKEPKPLEKSVSPQNSDSSGFADVNGWHLRFRWSKDAPSELLRKFRNYEI | |
| 100 μL | |
| Signal Transduction | |
| 56521 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction