Learn More
DNM1L Monoclonal Antibody (OTI1C1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA506116S
Encompass_Preferred
 View more versions of this product
Product Code. 17525472
  • 242.00€ / 30µL
Description
DNM1L Monoclonal antibody specifically detects DNM1L in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Specifications
Specifications
DNM1L
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
O00429, O35303, Q8K1M6
Mouse
Affinity chromatography
RUO
10059, 114114, 74006
-20° C, Avoid Freeze/Thaw Cycles
IgG2b
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
OTI1C1
Unconjugated
DNM1L
6330417M19Rik; AI450666; C-terminal region; Dlp1; dnm1l; Dnm1p/Vps1p-like protein; Dnml1; Dnmlp1; Drp1; DVLP; DYMPLE; dynamin 1 like; dynamin 1-like; dynamin family member proline-rich carboxyl-terminal domain less; Dynamin-1-like protein; Dynamin-1-like protein-like protein; Dynamin-like protein; dynamin-like protein 1; dynamin-like protein 4; Dynamin-like protein IV; dynamin-related protein 1; DYNIV 11; DYNIV-11; EC 3.6.5.5; EGK_03514; EMPF; FLJ41912; HDYNIV; N-terminal region; python; similar to dynamin 1-like; VPS1; zgc:66163
Full length human recombinant protein of human DNM1L produced in HEK293T cell.