missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dnmt2 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
176.00€ - 408.00€
Specifications
| Antigen | Dnmt2 |
|---|---|
| Dilution | Western Blot 1:1000, ELISA 1:100-1:1000, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:50-1:200, Immunohistochemistry-Frozen |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18365505
|
Novus Biologicals
NBP3-18663-25UL |
25 μg |
176.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18345095
|
Novus Biologicals
NBP3-18663-100UL |
100 μg |
408.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Dnmt2 Polyclonal antibody specifically detects Dnmt2 in Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)Specifications
| Dnmt2 | |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| Unconjugated | |
| Rabbit | |
| Chromatin Research, Epigenetics | |
| Whole antiserum | |
| DMNT2, DNA (cytosine-5-)-methyltransferase 2, DNA (cytosine-5)-methyltransferase-like protein 2, DNA methyltransferase homolog HsaIIP, DNA methyltransferase-2, DNA MTase homolog HsaIIP, Dnmt2, EC 2.1.1, EC 2.1.1.29, M.HsaIIP, MHSAIIP, PuMet, RNMT1, tRNA (cytosine-5-)-methyltransferase, tRNA aspartic acid methyltransferase 1 | |
| Polyclonal antibody raised in rabbit against mouse Dnmt2 (DNA methyltransferase 2), using a KLHconjugated synthetic peptide containing an amino acid sequence from the N-terminal part of the protein (1). | |
| Whole Antiserum |
| Western Blot 1:1000, ELISA 1:100-1:1000, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:50-1:200, Immunohistochemistry-Frozen | |
| Polyclonal | |
| Antisera | |
| RUO | |
| Mouse | |
| O14717 | |
| 1787 | |
| Primary | |
| Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title