missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNP Human HEK293, Unconjugated, Isotype Control, AcroBiosystems
Description
DNP Monoclonal antibody specifically detects DNP.
Specifications
Specifications
| Antigen | DNP |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Host Species | Human (HEK293) |
| Quantity | 100 μg |
| Regulatory Status | RUO |
| Content And Storage | -20°C |
| Isotype | IgG4 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?