missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ dNTPs Mix (25mm each)

Product Code. 11467120 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 μmol
5 x 5 μmol
Unit Size:
25µmol
5µmol
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11467120 5 μmol 5µmol
11477120 5 x 5 μmol 25µmol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11467120 Supplier MP Biomedicals™ Supplier No. 11NTPMX250
  • ONLINE EXCLUSIVE PRICE
    80.50€
    76.65€ / 5µmol
    Save 3.85€
    4% Off
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Improves PCR efficiency, lowers denaturation temperatures and increases template lengths. MP Biomedicals dNTPs Mix (25mm each) is a ready to use solution with guaranteed purification of dNTPs (>99% pure) in order to achieve sensitive and consistent res

Manufacturing process guarantees that the dNTPs are highly purified, as contaminants can hinder PCR and result in a disappointing product yield. This product achieves sensitive and consistent results with specially prepared deoxynucleotides at a concentration of 25mm.

It is suitable for use in a range of molecular biology applications, including: PCR, real-time PCR, high fidelity and long PCR, RT-PCR, cDNA synthesis, and DNA sequencing and labeling procedures.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 25 mM
Content And Storage dNTP mix contains 5 μmol of each dATP, dCTP, dGTP, dTTP. Store at -20°C.
pH 7
Purity or Quality Grade 99+%
Quantity 5 μmol
For Use With (Application) Standard PCR, Real Time PCR (qPCR), RT-PCR
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.