missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DOCK10 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16834-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the sorting nexin family. Members of this family contain a phox (PX) domain, which is a phosphoinositide binding domain, and are involved in intracellular trafficking. Overexpression of this gene results in a decrease in the processing of insulin and hepatocyte growth factor receptors to their mature subunits. This decrease is caused by the mislocalization of furin, the endoprotease responsible for cleavage of insulin and hepatocyte growth factor receptors. This protein is involved in endosomal trafficking from the plasma membrane to recycling endosomes or the trans-Golgi network. This gene encodes two transcript variants encoding distinct isoforms.Specifications
| DOCK10 | |
| Polyclonal | |
| Unconjugated | |
| DOCK10 | |
| dedicator of cytokinesis 10, DOCK10, DRIP2, KIAA0694, Nbla10300, ZIZ3, Zizimin 3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 210293, 301556, 55619 | |
| -20°C | |
| Liquid |
| Immunoprecipitation | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8BZN6, Q96BY6 | |
| DOCK10 | |
| DOCK10 Fusion Protein Ag10486 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction