missing translation for 'onlineSavingsMsg'
Learn More

Docosyl Ferulate, TRC

Product Code. 30384532
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30384532 10 mg
30380120 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30384532 Supplier Toronto Research Chemicals Supplier No. TRCD49465010MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101927-24-6
Molecular Formula C32 H54 O4
Molecular Weight (g/mol) 502.77
Synonym (E)-3-(4-Hydroxy-3-methoxyphenyl)-2-propenoic Acid Docosyl Ester, Docosyl trans-ferulate, E-Ferulic Acid Docosyl Ester
IUPAC Name docosyl (E)-3-(4-hydroxy-3-methoxyphenyl)prop-2-enoate
SMILES CCCCCCCCCCCCCCCCCCCCCCOC(=O)\C=C\c1ccc(O)c(OC)c1

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C32H54O4/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-27-36-32(34)26-24-29-23-25-30(33)31(28-29)35-2/h23-26,28,33H,3-22,27H2,1-2H3/b26-24+
Formula Weight 502.4022
Chemical Name or Material Docosyl Ferulate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.