Learn More
ZVG Zellstoff Vertrieb™ Doctors Tissue Role
Product Code. 10623303
Change view
Click to view available options
Dimensions (L x W):
50 m x 50 cm
Unit Size:
9 rolls
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
10623303 50 m x 50 cm 9 rolls
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10623303 Supplier ZVG Zellstoff Vertrieb™ Supplier No. 1222500
  • ONLINE EXCLUSIVE PRICE
    113.72€
    40.31€ / 9 rolls
    Save 73.41€
    64% Off
View all promotions
Estimated Shipment: 08-12-2026