missing translation for 'onlineSavingsMsg'
Learn More

Dodeca-Tungstosilicic Acid Hydrate, Extra Pure, SLR, Fisher Chemical

Product Code. 10529020 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Plastic powder jar
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10529020 100 g Plastic powder jar 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10529020 Supplier Fisher Chemical Supplier No. T/3940/48

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 12027-43-9
Color Yellow
Packaging Plastic powder jar
Molecular Formula H4O40SiW12
MDL Number MFCD00036271
Quantity 100 g
Synonym tungstosilicic acid, tungstosilicic acid hydrate, p.a., tungstosilicic acid hydrate, purum p.a., tungstosilicic acid hydrate, reagent grade, tungstosilicic acid hydrate trace metals basis, tungstate 4-, m12-orthosilicato 4--ko:ko:ko:ko':ko':ko':ko:ko:ko:ko':ko':ko' tetracosa-m-oxododecaoxododeca-, hydrogen 1:4
InChI Key HIKIIEGSHYIJCC-UHFFFAOYSA-K
SMILES O[Si](O[W](=O)(=O)O[W](=O)(=O)O[W](=O)(=O)O[W](O)(=O)=O)(O[W](=O)(=O)O[W](=O)(=O)O[W](=O)(=O)O[W](O)(=O)=O)O[W](=O)(=O)O[W](=O)(=O)O[W](=O)(=O)O[W](O)(=O)=O
IUPAC Name silicic acid;1,3,5-trioxa-2$l^{6},4$l^{6},6$l^{6}-tritungstacyclohexane 2,2,4,4,6,6-hexaoxide;hydrate
Molecular Weight (g/mol) 2878.16
PubChem CID 71307034
Physical Form Solid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.