missing translation for 'onlineSavingsMsg'
Learn More

Dodecanedioic Dihydrazide, 98%, BLD Pharm™

Product Code. 30984140
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30984140 5 g -
30976088 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30984140 Supplier BLD Pharm Supplier No. BD1267625g
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 4080-98-2
Molecular Formula C12H26N4O2
MDL Number MFCD00043619
Quantity 5 g
SMILES O=C(NN)CCCCCCCCCCC(NN)=O
Molecular Weight (g/mol) 258.36
Percent Purity 98%
Chemical Name or Material Dodecanedioic Dihydrazide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.