missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dodecyltrimethylammonium chloride 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| Chemical Name or Material | Dodecyltrimethylammonium Chloride |
| CAS | 112-00-5 |
| Analyte or Component Names | Dodecyltrimethylammonium Chloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C15 H34 N . Cl |
| Concentration | 100 μg/mL |
| Certifications/Compliance | ISO 17034 |
| InChI Formula | InChI=1S/C15H34N.ClH/c1-5-6-7-8-9-10-11-12-13-14-15-16(2,3)4;/h5-15H2,1-4H3;1H/q+1;/p-1 |
| SMILES | [Cl-].CCCCCCCCCCCC[N+](C)(C)C |
| IUPAC Name | dodecyl(trimethyl)azanium;chloride |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction