Learn More
DOK7 Monoclonal Antibody (OTI5G8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA504809S
Description
DOK7 Monoclonal antibody specifically detects DOK7 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western BlotSpecifications
| DOK7 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P58340 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 4291 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| OTI5G8 | |
| Unconjugated | |
| MLF1 | |
| A930013K19Rik; AW049091; C4orf25; CMS10; CMS1B; docking protein 7; DOK7; Dok-7; Downstream of tyrosine kinase 7; EF-12; Oit5; oncoprotein induced transcript 5; protein Dok-7; RGD1566416 | |
| Full length human recombinant protein of human MLF1 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.