missing translation for 'onlineSavingsMsg'
Learn More

Dolutegravir Sodium, TRC

Product Code. 17971645
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17971645 10 mg 10mg
17962945 100 mg 100mg
17934015 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17971645 Supplier Toronto Research Chemicals Supplier No. TRCD52880510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1051375-19-9
Molecular Formula C20 H18 F2 N3 O5 . Na
Molecular Weight (g/mol) 441.36
Synonym 2H-Pyrido[1',2':4,5]pyrazino[2,1-b][1,3]oxazine-9-carboxamide, N-[(2,4-difluorophenyl)methyl]-3,4,6,8,12,12a-hexahydro-7-hydroxy-4-methyl-6,8-dioxo-, sodium salt (1:1), (4R,12aS)-, Dolutegravir sodium, Dolutegravir sodium salt, GSK 1349572A, (4R,12aS)-N-[(2,4-Difluorophenyl)methyl]-3,4,6,8,12,12a-hexahydro-7-hydroxy-4-methyl-6,8-dioxo-2H-pyrido[1',2':4,5]pyrazino[2,1-b][1,3]oxazine-9-carboxamide sodium salt, (4R,9aS)-5-Hydroxy-4-methyl-6,10-dioxo-3,4,6,9,9a,10-hexahydro-2H-1-oxa-4a,8a-diaza-anthracene-7-carboxylic acid 2,4-difluoro-benzylamide sodium salt, 2H-Pyrido[1',2':4,5]pyrazino[2,1-b][1,3]oxazine-9-carboxamide, N-[(2,4-difluorophenyl)methyl]-3,4,6,8,12,12a-hexahydro-7-hydroxy-4-methyl-6,8-dioxo-, sodium salt (1:1), (4R,12aS)-, Dolutegravir sodium, Dolutegravir sodium salt, GSK 1349572A, GSK 1349572A
IUPAC Name sodium;(4R,12aS)-9-[(2,4-difluorophenyl)methylcarbamoyl]-4-methyl-6,8-dioxo-3,4,12,12a-tetrahydro-2H-pyrido[5,6]pyrazino[2,6-b][1,3]oxazin-7-olate
SMILES [Na+].C[C@@H]1CCO[C@H]2CN3C=C(C(=O)NCc4ccc(F)cc4F)C(=O)C(=C3C(=O)N12)[O-]

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C20H19F2N3O5.Na/c1-10-4-5-30-15-9-24-8-13(17(26)18(27)16(24)20(29)25(10)15)19(28)23-7-11-2-3-12(21)6-14(11)22;/h2-3,6,8,10,15,27H,4-5,7,9H2,1H3,(H,23,28);/q;+1/p-1/t10-,15+;/m1./s1
Formula Weight 441.1112
Percent Purity >95%
Chemical Name or Material Dolutegravir Sodium Salt
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.