Learn More
Dopa Decarboxylase/DDC Antibody (CL2962) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-46642JF525
Product Code. 30295502
  • 737.00€ / 0.10mL
Description
Dopa Decarboxylase/DDC Monoclonal antibody specifically detects Dopa Decarboxylase/DDC in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Dopa Decarboxylase/DDC
Monoclonal
Janelia Fluor 525
AADCaromatic-L-amino-acid decarboxylase, DOPA decarboxylase, dopa decarboxylase (aromatic L-amino acid decarboxylase), EC 4.1.1, EC 4.1.1.28
This antibody was developed using a recombinant protein derived from P20711, with the exact immunogen sequence remaining proprietary.
0.1 mL
Neuroscience
1644
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
CL2962
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human, Mouse, Rat