missing translation for 'onlineSavingsMsg'
Learn More

Dopamine 3-O-Sulfate, TRC

Product Code. 30009407
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30009407 50 mg 50mg
30005532 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30009407 Supplier Toronto Research Chemicals Supplier No. TRCD53384050MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51317-41-0
Molecular Formula C8 H11 N O5 S
Molecular Weight (g/mol) 233.24
Synonym Pyrocatechol, 4-(2-aminoethyl)-, 2-(hydrogen sulfate) (7CI), Dopamine 3-O-sulfate, Dopamine 3-sulfate, Dopamine sulfate, [5-(2-Aminoethyl)-2-hydroxyphenyl]oxidanesulfonic acid, 1,2-Benzenediol, 4-(2-aminoethyl)-, 2-(hydrogen sulfate), Dopamine 3-O-Sulfate
IUPAC Name [5-(2-aminoethyl)-2-hydroxyphenyl] hydrogen sulfate
SMILES NCCc1ccc(O)c(OS(=O)(=O)O)c1

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C8H11NO5S/c9-4-3-6-1-2-7(10)8(5-6)14-15(11,12)13/h1-2,5,10H,3-4,9H2,(H,11,12,13)
Formula Weight 233.0358
Percent Purity >95%
Chemical Name or Material Dopamine 3-O-Sulfate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.