Learn More
Invitrogen™ Dostarlimab Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA559247
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Dostarlimab (TSR-042) is a humanized anti-PD-1 monoclonal antibody jointly developed by TESARO and AnaptysBio. It binds to the PD-1 receptor with high affinity, thereby blocking its binding to PD-L1 and PD-L2 ligands.
Specifications
| Dostarlimab Humanized | |
| Recombinant Monoclonal | |
| Unconjugated | |
| ANB-011; TSR-042 | |
| Protein A | |
| RUO | |
| Human | |
| Antibody | |
| IgG4SP |
| ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance | |
| 1 mg/mL | |
| 25 mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 | |
| Human | |
| 100 μg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |