missing translation for 'onlineSavingsMsg'
Learn More

DOTP, TRC

Product Code. 30385700
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30385700 1 g
30383009 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30385700 Supplier Toronto Research Chemicals Supplier No. TRCD4948901G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91987-74-5
Molecular Formula C12H32N4O12P4
Molecular Weight (g/mol) 548.3
Synonym 1,4,7,10-Tetraazacyclododecane-1,4,7,10-tetra(methylene Phosphonic Acid), 1,4,7,10-Tetraazacyclododecane, Phosphonic Acid deriv., 1,4,7,10-Tetraazacyclododecane-1,4,7,10-tetramethylenephosphonic Acid, DOTMP, NSC 681106, N,N',N'',N'''-Tetrakis(phosphonomethyl)-1,4,7,10-tetraazacyclododecane
SMILES OP(=O)(O)CN1CCN(CP(=O)(O)O)CCN(CP(=O)(O)O)CCN(CP(=O)(O)O)CC1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C12H32N4O12P4/c17-29(18,19)9-13-1-2-14(10-30(20,21)22)5-6-16(12-32(26,27)28)8-7-15(4-3-13)11-31(23,24)25/h1-12H2,(H2,17,18,19)(H2,20,21,22)(H2,23,24,25)(H2,26,27,28)
Chemical Name or Material DOTP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.