Learn More
Azpack™ Sieve Cleaning Brush
Product Code. 12358069
Change view
Click to view available options
Color:
Black, White
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
12358069 Black, White Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12358069 Supplier Azpack™ Supplier No. 12358069
  • ONLINE EXCLUSIVE PRICE
    16.00€
    10.59€ / Pack of 1
    Save 5.41€
    33% Off
Estimated Shipment: 23-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Remove stubborn materials with the stiff side, dust particles away with the soft side. Azpack™ Sieve Cleaning Brush is dual-ended for optimum cleaning efficiency.
  • Soft nylon bristles on one end, stiff nylon bristles on the opposite end.
  • Flat bristles are black on one end, white on the other
  • Wooden handle with metal ferrules
Specifications
Product Type Brush
Handle Material Wood
Color Black, White
Length (English) 9.25 in.
Length (Metric) 235 mm
Width (English) Bristle 0.74 in.
Width (Metric) Bristle 19 mm