missing translation for 'onlineSavingsMsg'
Learn More

Doxazosin Mesilate EP Impurity C (Doxazosin USP Related Compound F), Mikromol™

Product Code. 30052041
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30052041 25 mg 25mg
30049162 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30052041 Supplier Mikromol Supplier No. MM0386.060025
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Doxazosin Mesilate EP Impurity C (Doxazosin USP Related Compound F)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,4-Bis(2,3-dihydro-1,4-benzodioxin-2-ylcarbonyl)piperazine
CAS 617677-53-9
Analyte or Component Names 1,4-Bis(2,3-dihydro-1,4-benzodioxin-2-ylcarbonyl)piperazine
Shipping Condition Room Temperature
Molecular Formula C22 H22 N2 O6
Certifications/Compliance ISO 17025
Synonym Doxazosin Mesilate Imp. C (EP), Doxazosin USP Related Compound F, Doxazosin USP RC F, 1,4-Bis(2,3-dihydro-1,4-benzodioxin-2-ylcarbonyl)piperazine, Doxazosin Related Compound F, Doxazosin Mesilate Impurity C, Doxazosin Impurity C
InChI Formula InChI=1S/C22H22N2O6/c25-21(19-13-27-15-5-1-3-7-17(15)29-19)23-9-11-24(12-10-23)22(26)20-14-28-16-6-2-4-8-18(16)30-20/h1-8,19-20H,9-14H2
SMILES O=C(C1COc2ccccc2O1)N3CCN(CC3)C(=O)C4COc5ccccc5O4
IUPAC Name [4-(2,3-dihydro-1,4-benzodioxine-3-carbonyl)piperazin-1-yl]-(2,3-dihydro-1,4-benzodioxin-3-yl)methanone
Molecular Weight (g/mol) 410.42
Quantity 25 mg
Formula Weight 410.1478 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.