missing translation for 'onlineSavingsMsg'
Learn More

Doxepin Hydrochloride, LoGiCal

Product Code. 30461944
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30461944 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30461944 Supplier LoGiCal Supplier No. LGCFOR0032.00

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Doxepin Hydrochloride
CAS 1229-29-4
Analyte or Component Names Doxepin Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C19 H21 N O . Cl H
Certifications/Compliance ISO 9001
Synonym Doxepin Hydrochloride, 3-Dibenz[b,e]oxepin-11(6H)-ylidene-N,N-dimethyl-1-propanamine hydrochloride, Dibenz[b,e]oxepin, 1-propanamine deriv., Adapin, Aponal, Curatin, Doxepin hydrochloride, Doxepine hydrochloride, P 3693A, Quitaxon, Sinequan, Sinequin, N,N-Dimethyldibenz[b,e]oxepin-Delta11(6H),gamma-propylamine hydrochloride
InChI Formula InChI=1S/C19H21NO.ClH/c1-20(2)13-7-11-17-16-9-4-3-8-15(16)14-21-19-12-6-5-10-18(17)19;/h3-6,8-12H,7,13-14H2,1-2H3;1H/b17-11+;
Solution Type Neat
SMILES Cl.CN(C)CC\C=C\1/c2ccccc2COc3ccccc13
IUPAC Name (3E)-3-(6H-benzo[c][1]benzoxepin-11-ylidene)-N,N-dimethylpropan-1-amine;hydrochloride
Industry Type Clinical Testing
Molecular Weight (g/mol) 315.84
Quantity 10 mg
Formula Weight 315.139 g/mol
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.