missing translation for 'onlineSavingsMsg'
Learn More

Doxepin hydrochloride, Thermo Scientific™

Potent H1-histamine receptor antagonist and a 5-HT receptor antagonist

Brand:  Thermo Scientific Alfa Aesar J62579.03

Quantity 1g

 View more versions of this product

Product Code. 15494919

  • 46.50€ / 1g
 Restricted Item
In Stock
This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Solubility
Soluble in water (100 mM), chloroform (1:2), alcohol (1:1), methanol, and DMSO.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

1229-29-4
315.841
MHNSPTUQQIYJOT-SJDTYFKWSA-N
6419921
CN(C)CCC=C1C2=CC=CC=C2COC3=CC=CC=C31.Cl
C19H22ClNO
MFCD00079135
doxepin hydrochloride, novoxapin, aponal, toruan, curatin, sinequan, zonalon, adapin, doxepine hydrochloride, novoxapin hydrochloride
(3E)-3-(6H-benzo[c][1]benzoxepin-11-ylidene)-N,N-dimethylpropan-1-amine;hydrochloride

Specifications

1229-29-4
MFCD00079135
UN2811
doxepin hydrochloride, novoxapin, aponal, toruan, curatin, sinequan, zonalon, adapin, doxepine hydrochloride, novoxapin hydrochloride
MHNSPTUQQIYJOT-SJDTYFKWSA-N
(3E)-3-(6H-benzo[c][1]benzoxepin-11-ylidene)-N,N-dimethylpropan-1-amine;hydrochloride
6419921
Doxepin hydrochloride
C19H22ClNO
1g
14,3435
Soluble in water (100 mM),chloroform (1:2),alcohol (1:1),methanol,and DMSO.
CN(C)CCC=C1C2=CC=CC=C2COC3=CC=CC=C31.Cl
315.841
315.84
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.