missing translation for 'onlineSavingsMsg'
Learn More

(±)-Doxylamine, TRC

Product Code. 30136467
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
Unit Size:
1g
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136467 1 g 1g
30003853 500 mg 500mg
30005102 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136467 Supplier Toronto Research Chemicals Supplier No. TRCD5620051G
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 469-21-6
Molecular Formula C17H22N2O
Molecular Weight (g/mol) 270.37
Synonym N,N-Dimethyl-2-[1-phenyl-1-(2-pyridinyl)ethoxy]ethanamine, 2-[alpha-[2-(Dimethylamino)ethoxy]-alpha-methylbenzyl]pyridine, Doxylamine, LS 127;
SMILES CN(C)CCOC(C)(c1ccccc1)c2ccccn2

Specifications

Quantity 1 g
InChI Formula InChI=1S/C17H22N2O/c1-17(20-14-13-19(2)3,15-9-5-4-6-10-15)16-11-7-8-12-18-16/h4-12H,13-14H2,1-3H3
Formula Weight 270.1732
Percent Purity >95%
Chemical Name or Material (±)-Doxylamine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.