missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DPCR-1 Antibody (213201) [DyLight 350], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB3347D
This item is not returnable.
View return policy
Description
DPCR-1 Monoclonal antibody specifically detects DPCR-1 in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
| DPCR-1 | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 135656 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Flow Cytometry, CyTOF | |
| 213201 | |
| Flow Cytometry, CyTOF-ready | |
| bCX105N19.6, C6orf37, diffuse panbronchiolitis critical region 1, MGC126710, MGC126712, PBLTdiffuse panbronchiolitis critical region 1 protein | |
| NS0 mouse myeloma cell line transfected with human DPCR-1, Met1-Arg235, Accession # AAH69477 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction