missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DPRX Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA570783
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Human: 100%.
Homeobox genes encode DNA-binding proteins, many of which are thought to be involved in early embryonic development. Homeobox genes encode a DNA-binding domain of 60 to 63 amino acids referred to as the homeodomain. This gene is a member of the DPRX homeobox gene family. Evidence of mRNA expression has not yet been found for this gene. Multiple, related processed pseudogenes have been found which are thought to reflect expression of this gene in the germ line or embryonic cells.
Specifications
| DPRX | |
| Polyclonal | |
| Unconjugated | |
| DPRX | |
| Divergent paired-related homeobox; divergent-paired related homeobox; DPRX | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 503834 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| A6NFQ7 | |
| DPRX | |
| synthetic peptide directed towards the N-terminal region of human DPRX. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto