Learn More
DPY30 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00€ - 410.00€
Specifications
| Antigen | DPY30 |
|---|---|
| Concentration | 400 μg/mL |
| Applications | Immunohistochemistry, Immunofluorescence, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17264606
|
Proteintech
16281-1-AP-20UL |
20 μL |
145.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17254606
|
Proteintech
16281-1-AP-150UL |
150 μL |
410.00€
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The DPY30 antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human DPY30. This antibody recognizes human, mouse, rat antigen. The DPY30 antibody has been validated for the following applications: IF, IHC,ELISA analysis.
Specifications
| DPY30 | |
| Immunohistochemistry, Immunofluorescence, ELISA | |
| Unconjugated | |
| Rabbit | |
| 84661 | |
| Primary |
| 400 μg/mL | |
| Polyclonal | |
| Liquid | |
| Human, Mouse, Rat | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.