Learn More
Dräger™ CMS Chips: Carbon Dioxide
Product Code. 12314143
Change view
Click to view available options
Measurement Range:
1 to 20%
1000 to 25000
200 to 3000
Response Time:
12 to 120 sec.
25 to 140 sec.
60 to 260 sec.
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Measurement Range Response Time Unit Size
12314143 1 to 20% 12 to 120 sec. Each
10669285 200 to 3000 60 to 260 sec. Each
10573612 1000 to 25000 25 to 140 sec. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12314143 Supplier Dräger™ Supplier No. 6406210
  • ONLINE EXCLUSIVE PRICE
    242.82€
    196.80€ / Pack of 1
    Save 46.02€
    18% Off
Estimated Shipment: 11-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Suitable for 10 measurements of one gas
 Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.
Specifications
Air Pressure 700 to 1100 hpa
Type Carbon Dioxide CMS Chip
Humidity Range 1 to 30mg/L
Measurement Range 1 to 20%
Response Time 12 to 120 sec.
Temperature Range (English) 32°F to 104°F
Temperature Range (Metric) 0°C to 40°C