Learn More
Dräger™ CMS Chips: Petroleum Hydrocarbons
Product Code. 12324639
Change view
Click to view available options
Measurement Range:
100 to 3000
20 to 500
Response Time:
150 to 330 sec.
30 to 110 sec.
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Measurement Range Response Time Unit Size
12324639 100 to 3000 30 to 110 sec. Each
10446301 20 to 500 150 to 330 sec. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12324639 Supplier Dräger™ Supplier No. 6406270
  • ONLINE EXCLUSIVE PRICE
    252.41€
    200.56€ / Pack of 1
    Save 51.85€
    20% Off
Estimated Shipment: 11-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Suitable for 10 measurements of one gas
Automatically supplies the analyzer with the calibration data and measurement range of the respective gas.
Specifications
Air Pressure 700 to 1100 hpa
Type Petroleum Hydrocarbons CMS Chip
Humidity Range 1 to 30mg/L
Measurement Range 100 to 3000
Response Time 30 to 110 sec.
Temperature Range (English) 32°F to 104°F
Temperature Range (Metric) 0°C to 40°C