Learn More
Dräger™ Short-Term Detector Tubes: Benzene
Product Code. 10443291
Change view
Click to view available options
Measurement Range:
0.5 to 10
15 to 420
2 to 60
5 to 50
Tube Type:
Benzene 0.5/a
Benzene 0.5/c
Benzene 15/a
Benzene 2/a
Benzene 5/b
Humidity Range:
1 to 15mg/L
1 to 30mg/L
3 to 15mg/L
Unit Size:
Each
Pack of 10
Pack of 5
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Measurement Range Tube Type Humidity Range unitSize
10443291 15 to 420 Benzene 15/a 1 to 30mg/L Each
10553802 0.5 to 10 Benzene 0.5/a 3 to 15mg/L Pack of 10
10430102 2 to 60 Benzene 2/a 1 to 15mg/L Pack of 5
12304539 5 to 50 Benzene 5/b 3 to 15mg/L Pack of 10
10743263 0.5 to 10 Benzene 0.5/c 1 to 30mg/L Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 10443291 Supplier Dräger™ Supplier No. 8101741
  • ONLINE EXCLUSIVE PRICE
    182.00€
    150.68€ / Pack of 1
    Save 31.32€
    17% Off
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
Detector tubes
Glass detector tubes are filled with an inert reagent carrier material (silica gel, aluminum oxide, or aluminum silicate) impregnated with an indicating reagent. Reagent produces a colorimetric indication in the presence of a particular gas, vapor, or aerosol. Concentrations can be read directly from the color change on the tube's printed scale. Tubes are hermetically sealed. Most have a two-year shelf life (when stored at room temperature). Available in packs of ten. Note: Some tests require a pretube or comparison tube in addition to the detector tube. These tubes are included in the box of 10, thus reducing the number of detector tubes. The actual number of detector tubes is cited in the ordering charts.
Order Info
Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.
Specifications
Components Measured Benzene
Humidity Range 1 to 30mg/L
Measurement Range 15 to 420
No. of Strokes 20 to 2
Quantity 10
Response Time 4 min.
Temperature Range (English) 32°F to 86°F
Temperature Range (Metric) 0°C to 30°C
Tube Type Benzene 15/a