Learn More
Dräger™ Short-Term Detector Tubes: Formaldehyde
Product Code. 12712517
Change view
Click to view available options
Measurement Range:
0.2 to 5
2 to 40
Tube Type:
Formaldehyde 0.2/a
Formaldehyde 2/a
Unit Size:
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Measurement Range Tube Type Unit Size
12712517 2 to 40 Formaldehyde 2/a Pack of 10
10059050 0.2 to 5 Formaldehyde 0.2/a Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12712517 Supplier Dräger™ Supplier No. 8101751
  • ONLINE EXCLUSIVE PRICE
    298.20€
    231.19€ / Pack of 10
    Save 67.01€
    22% Off
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Detector tubes
Glass detector tubes are filled with an inert reagent carrier material (silica gel, aluminum oxide, or aluminum silicate) impregnated with an indicating reagent. Reagent produces a colorimetric indication in the presence of a particular gas, vapor, or aerosol. Concentrations can be read directly from the color change on the tube's printed scale. Tubes are hermetically sealed. Most have a two-year shelf life (when stored at room temperature). Available in packs of ten.
Note: Some tests require a pretube or comparison tube in addition to the detector tube. These tubes are included in the box of 10, thus reducing the number of detector tubes. The actual number of detector tubes is cited in the ordering charts.
Order Info
Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.
Specifications
Components Measured Formaldehyde
Type Gas Detection Tube
Humidity Range 3 to 15mg/L
Measurement Range 2 to 40
No. of Strokes 5
Quantity 10
Response Time 30 sec.
Temperature Range (English) 32°F to 104°F
Temperature Range (Metric) 0°C to 40°C
Tube Type Formaldehyde 2/a