Learn More
Dräger™ Short-Term Detector Tubes - De Tube Hydrochloric Acid
Description
- Glass vial
- Calibrated 100mL sample of air drawn through tube with Drager Accuro™ bellows pump
- Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.
- Safety Equipment Institute (SEI) certified.
Specifications
Specifications
| Components Measured | Hydrochloric Acid 50/a |
| For Use With (Equipment) | Accuro Drager Gas Detector Pump |
| Type | Gas Detection Tube |
| Humidity Range | 15mg/L |
| Measurement Range | 50 to 5000ppm |
| No. of Strokes | 1/10 |
| Quantity | 10 |
| Response Time | 5 min./30 sec. |
| Temperature Range (English) | 50°F to 122°F |
| Temperature Range (Metric) | 10° to 50°C |