Learn More
Novus Biologicals™ DR6/TNFRSF21 Antibody (Abbvie patent anti-TNFRSF21) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-28486-100ug
 View more versions of this product
Product Code. 30045559
  • 338.00€ / 100µg
Description
DR6/TNFRSF21 Monoclonal antibody specifically detects DR6/TNFRSF21 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
DR6/TNFRSF21
Monoclonal
Unconjugated
O75509
Human
Protein A purified
RUO
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Store at -20°C in powder form. Store at -80°C once reconstituted.
IgG1
Flow Cytometry, ELISA, Functional Assay
Abbvie patent anti-TNFRSF21
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
BM-018, CD358 antigen, Death receptor 6, DR6TNFR-related death receptor 6, MGC31965, tumor necrosis factor receptor superfamily member 21, tumor necrosis factor receptor superfamily, member 21
TNFRSF21/ DR6/ CD358
100 μg
Apoptosis, Cell Biology
27242