missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DRAK1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
189.00€
Specifications
| Antigen | DRAK1 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 2-10 μg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
DRAK1 Polyclonal specifically detects DRAK1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| DRAK1 | |
| Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 2-10 μg/mL | |
| Polyclonal | |
| Rabbit | |
| Apoptosis, Cell Biology, Signal Transduction | |
| PBS with 0.02% Sodium Azide | |
| DAP kinase-related apoptosis-inducing protein kinase 1, DRAK1death-associated protein kinase-related 1, EC 2.7.11.1, serine/threonine kinase 17a, serine/threonine kinase 17a (apoptosis-inducing), serine/threonine-protein kinase 17A | |
| STK17A | |
| IgG | |
| Affinity Purified | |
| No cross responses to DRAK2, DAP or ZIP kinases. |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| Q9UEE5 | |
| 9263 | |
| Antibody was raised against a peptide corresponding to amino acids near the amino terminus of human DRAK1. The immunogen is located within the first 50 amino acids of DRAK1. Amino Acid Squence: EKPGSGGSSPGATSG | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| 50 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title