missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DRAM Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
178.00€
Specifications
| Antigen | DRAM |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1-2 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 2-2.5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 2-2.5 μg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
DRAM Polyclonal specifically detects DRAM in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunofluorescence.Specifications
| DRAM | |
| Western Blot 1-2 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 2-2.5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 2-2.5 μg/mL | |
| Polyclonal | |
| Rabbit | |
| Cancer | |
| PBS with 0.02% Sodium Azide | |
| Damage-regulated autophagy modulator, DNA damage-regulated autophagy modulator protein 1, DNA-damage regulated autophagy modulator 1, DRAMFLJ11259 | |
| DRAM1 | |
| IgG | |
| Affinity Purified | |
| 30 kDa |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| AAH18435 | |
| 55332 | |
| Antibody was raised against a 16 amino acid synthetic peptide from near the amino terminus of human DRAM. The immunogen is located within amino acids 30 - 80 of DRAM. Amino Acid Squence: LPYISDTGTTPPESGI | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title