missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ DRI Tricyclics Serum Toxicology Assays

Product Code. 15689275 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
25 x 8 mL
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15689275 25 x 8 mL 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15689275 Supplier Thermo Scientific™ Supplier No. 1128
  • ONLINE EXCLUSIVE PRICE
    312.00€
    278.07€ / 1 sets
    Save 33.93€
    10% Off
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Quickly and accurately identify tricyclics in serum samples with Thermo Scientific™ DRI™ Tricyclics Serum Toxicology Assays, excellent tools for emergency rooms when urine specimens are not conveniently available.

These Serum Toxicology Tricyclics Assays have the flexibility of analyzing serum or urine samples. These assays are provided in convenient packaging with separate calibrators for ease of use. A multi-analyte Serum Tox Control is also available. The assays are liquid, ready-to-use and have the versatility to be run on a variety of clinical chemistry analyzers.
  • Serum-and urine-based assay
  • Used to identify class of drug overdose

Ordering Alerts:

1128: Reagent volume 25, 8mL.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Description DRI Tricyclics
Detectable Analytes Tricyclic Antidepressants (TCA)
Sample Type Serum
Type Reagent
Quantity 25 x 8 mL

For sale in the U.S.A. and Canada only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.