missing translation for 'onlineSavingsMsg'
Learn More

Droxicam, TRC

Product Code. 30134068
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134068 500 mg 500mg
30138688 1 g 1g
30007466 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134068 Supplier Toronto Research Chemicals Supplier No. TRCD689880500MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90101-16-9
Molecular Formula C16 H11 N3 O5 S
Molecular Weight (g/mol) 357.34
Synonym 2H,5H-1,3-Oxazino[5,6-c][1,2]benzothiazine-2,4(3H)-dione, 5-methyl-3-(2-pyridinyl)-, 6,6-dioxide, Dobenam, Droxar, Droxicam, E 3128, E 318, Ombolan
IUPAC Name 5-methyl-6,6-dioxo-3-pyridin-2-yl-[1,3]oxazino[5,6-c][1,2]benzothiazine-2,4-dione
SMILES CN1C2=C(OC(=O)N(C2=O)c3ccccn3)c4ccccc4S1(=O)=O

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C16H11N3O5S/c1-18-13-14(10-6-2-3-7-11(10)25(18,22)23)24-16(21)19(15(13)20)12-8-4-5-9-17-12/h2-9H,1H3
Formula Weight 357.0419
Chemical Name or Material Droxicam
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.