missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ DS2

Description
DS2 is a positive allosteric modulator of GABAA receptors. Potentiates GABA-evoked currents mediated by α4β3δ receptors (EC50 = 142 nM, in vitro). Displays complex selectivity profile, with selectivity for δ-containing receptors (α4/6βxδ > α1βxδ >> γ2 > α4β3 GABAA receptors). Enhances tonic currents of thalamocortical neurons in wild-type mice.
Specifications
Specifications
| Quantity | 10 mg |
| Product Type | DS2 |
| Purity | >97% |
| Molecular Weight (g/mol) | 353.83 |
| Inhibitors | DS2 |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?