missing translation for 'onlineSavingsMsg'
Learn More
Learn More
dsDNA Antibody (DSD/8204R), Alexa Fluor™ 700, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-20860AF700
This item is not returnable.
View return policy
Description
dsDNA Monoclonal antibody specifically detects dsDNA in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| dsDNA | |
| Monoclonal | |
| Alexa Fluor 700 | |
| Double stranded DNA | |
| Nuclei of Burkitt's cells | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| DSD/8204R | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction